missing translation for 'onlineSavingsMsg'
Learn More

6-Iodo Diosmin (1 mg/mL in DMSO), TRC

Product Code. 30390959
Change view
Click to view available options
Quantity:
1x1 mL
Unit Size:
1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30390959 1x1 mL 1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30390959 Supplier Toronto Research Chemicals Supplier No. TRCKIT07351X1ML
 Restricted Item
Estimated Shipment: 21-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 6-Iodo Diosmin
CAS 1431536-92-3
Molecular Formula C28 H31 I O15
Quantity 1x1 mL
Synonym 7-[[6-O-(6-Deoxy-α-L-mannopyranosyl)-β-D-glucopyranosyl]oxy]-5-hydroxy-2-(3-hydroxy-4-methoxyphenyl)-6-iodo-4H-1-benzopyran-4-one, 6-Iododiosmin, Diosmin Imp. D (EP)
InChI Formula InChI=1S/C28H31IO15/c1-9-20(32)23(35)25(37)27(41-9)40-8-17-21(33)24(36)26(38)28(44-17)43-16-7-15-18(22(34)19(16)29)12(31)6-14(42-15)10-3-4-13(39-2)11(30)5-10/h3-7,9,17,20-21,23-28,30,32-38H,8H2,1-2H3/t9-,17+,20-,21+,23+,24-,25+,26+,27+,28+/m0/s1
SMILES COc1ccc(cc1O)C2=CC(=O)c3c(O)c(I)c(O[C@@H]4O[C@H](CO[C@@H]5O[C@@H](C)[C@H](O)[C@@H](O)[C@H]5O)[C@@H](O)[C@H](O)[C@H]4O)cc3O2
IUPAC Name 5-hydroxy-2-(3-hydroxy-4-methoxyphenyl)-6-iodo-7-[(2S,3R,4S,5S,6R)-3,4,5-trihydroxy-6-[[(2R,3R,4R,5R,6S)-3,4,5-trihydroxy-6-methyloxan-2-yl]oxymethyl]oxan-2-yl]oxychromen-4-one
Molecular Weight (g/mol) 734.44
Formula Weight 734.0708
Concentration or Composition (by Analyte or Components) 1 mg/mL
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.