missing translation for 'onlineSavingsMsg'
Learn More

6-Iodo-2,3-dihydro-1,4-benzodioxine, 97%, Thermo Scientific™

Product Code. 10794763
Click to view available options
Amber Glass Bottle; 1g
This item is not returnable. View return policy
This item is not returnable. View return policy

Specifications

CAS 57744-67-9
CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Molecular Formula C8H7IO2
MDL Number MFCD00221460
Quantity 1g
Synonym 3,4-ethylenedioxyiodobenzene, 6-iodo-2,3-dihydrobenzo b 1,4 dioxine, 6-iodobenzodioxane, acmc-1azse, 6-iodo-1,4-benzodioxane, 6-iodo-2h,3h-benzo e 1,4-dioxin, 2,3-dihydro-6-iodobenzo 1,4 dioxin, 1,4-benzodioxin,2,3-dihydro-6-iodo
InChI Key HZTMYTXWFHBHDC-UHFFFAOYSA-N
SMILES IC1=CC=C2OCCOC2=C1
IUPAC Name 6-iodo-2,3-dihydro-1,4-benzodioxine
Molecular Weight (g/mol) 262.05
PubChem CID 2776176
Formula Weight 262.04
Percent Purity 97%
Chemical Name or Material 6-Iodo-2, 3-dihydro-1, 4-benzodioxine
Show More Show Less
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'