missing translation for 'onlineSavingsMsg'
Learn More

6-Hydroxytryptamine Creatinine Sulfate, TRC

Product Code. 30392544
Change view
Click to view available options
Quantity:
5 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30392544 5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30392544 Supplier Toronto Research Chemicals Supplier No. TRCH9750005MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

CAS 39929-25-4
Molecular Formula C10 H12 N2 O . C4 H9 N3 O2 . H2 O4 S
Quantity 5 mg
Synonym 4H-Imidazol-4-one, 2-amino-1,5-dihydro-1-methyl-, compd. with 3-(2-aminoethyl)-1H-indol-6-ol and sulfuric acid (1:1:1) (ACI), 1H-Indol-6-ol, 3-(2-aminoethyl)-, compd. with 2-amino-1,5-dihydro-1-methyl-4H-imidazol-4-one sulfate (salt) (1:1:1) (9CI), 4H-Imidazol-4-one, 2-amino-1,5-dihydro-1-methyl-, compd. with 3-(2-aminoethyl)-1H-indol-6-ol sulfate (salt) (1:1:1) (9CI), Indol-6-ol, 3-(2-aminoethyl)-, compd. with creatinine sulfate (1:1) (7CI), 6-Hydroxytryptamine creatinine sulfate
InChI Formula InChI=1S/C10H12N2O.C4H9N3O2.H2O4S/c11-4-3-7-6-12-10-5-8(13)1-2-9(7)10;1-7(4(5)6)2-3(8)9;1-5(2,3)4/h1-2,5-6,12-13H,3-4,11H2;2H2,1H3,(H3,5,6)(H,8,9);(H2,1,2,3,4)
SMILES CN(CC(=O)O)C(=N)N.NCCc1c[nH]c2cc(O)ccc12.OS(=O)(=O)O
IUPAC Name 3-(2-aminoethyl)-1H-indol-6-ol;2-[carbamimidoyl(methyl)amino]acetic acid;sulfuric acid
Molecular Weight (g/mol) 405.43
Formula Weight 405.1318
Chemical Name or Material 6-Hydroxytryptamine Creatinine Sulfate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.