missing translation for 'onlineSavingsMsg'
Learn More

6-Hydroxyquinoline-(1H)-2-one, TRC

Product Code. 30390896
Change view
Click to view available options
Quantity:
250 mg
1 g
2.5 g
Unit Size:
2.5g
250mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30390896 1 g -
30397618 250 mg 250mg
30394137 2.5 g 2.5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30390896 Supplier Toronto Research Chemicals Supplier No. TRCH9532501G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 19315-93-6
Molecular Formula C9 H7 N O2
Molecular Weight (g/mol) 161.16
Synonym 2(1H)-Quinolinone, 6-hydroxy-, 2,6-Quinolinediol (7CI,8CI), 6-Hydroxy-2(1H)-quinolinone, 2,6-Dihydroxyquinoline, 6-Hydroxy-1H-quinolin-2-one, 6-Hydroxycarbostyril, 6-Hydroxyquinolin-2-one
IUPAC Name quinoline-2,6-diol
SMILES Oc1ccc2nc(O)ccc2c1

Specifications

Quantity 1 g
InChI Formula InChI=1S/C9H7NO2/c11-7-2-3-8-6(5-7)1-4-9(12)10-8/h1-5,11H,(H,10,12)
Formula Weight 161.0477
Chemical Name or Material 6-Hydroxyquinoline-(1H)-2-one
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.