missing translation for 'onlineSavingsMsg'
Learn More

6-Hydroxypyridine-3-boronic acid pinacol ester, 97%

Product Code. 15459245
Change view
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15459245 250 mg 250mg
15449245 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15459245 Supplier Thermo Scientific Alfa Aesar Supplier No. H51673.MD
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 1054483-78-1 | C11H16BNO3 | 221.06 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1054483-78-1
Molecular Formula C11H16BNO3
Molecular Weight (g/mol) 221.06
MDL Number MFCD06795671
InChI Key WAUWXCUPDOXYKS-UHFFFAOYSA-N
Synonym 5-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl pyridin-2-ol, 6-hydroxypyridine-3-boronic acid pinacol ester, 2-hydroxypyridine-5-boronic acid pinacol ester, 2-hydroxypyridine-5-boronic acid, pinacol ester, 5-tetramethyl-1,3,2-dioxaborolan-2-yl pyridin-2-ol, 6-hydroxypyridine-3-boronic acid, pinacol ester, 5-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl pyridin-2 1h-one, 2-hydroxy-5-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl pyridine, amtb145, pyridin-2-ol-5-boronic acid pinacol ester
PubChem CID 17750253
IUPAC Name 5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-1H-pyridin-2-one
SMILES CC1(C)OB(OC1(C)C)C1=CN=C(O)C=C1

Specifications

Quantity 250 mg
Formula Weight 221.06
Percent Purity 97%
Chemical Name or Material 6-Hydroxypyridine-3-boronic acid pinacol ester

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.