missing translation for 'onlineSavingsMsg'
Learn More

6-Hydroxynicotinic acid, 98%

Product Code. 11429190
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11429190 100 g 100g
11409190 5 g 5g
11419190 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11429190 Supplier Thermo Scientific Alfa Aesar Supplier No. A15802.22
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 5006-66-6 | C6H5NO3 | 139.11 g/mol

Metabolite of nicotinamide. Stimulates adrenocortical secretion

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 5006-66-6
Molecular Formula C6H5NO3
Molecular Weight (g/mol) 139.11
MDL Number MFCD00006277
InChI Key BLHCMGRVFXRYRN-UHFFFAOYSA-N
Synonym 6-hydroxynicotinic acid, 2-hydroxy-5-pyridinecarboxylic acid, 6-hydroxypyridine-3-carboxylic acid, 6-hydroxy nicotinic acid, 3-pyridinecarboxylic acid, 1,6-dihydro-6-oxo, 2-hydroxypyridine-5-carboxylic acid, 2-pyridone-5-carboxylic acid, 6-hydroxy-nicotinic acid, 6-hydroxynicotinate, 1,6-dihydro-6-oxo-3-pyridinecarboxylic acid
PubChem CID 72924
ChEBI CHEBI:16168
IUPAC Name 6-oxo-1H-pyridine-3-carboxylic acid
SMILES C1=CC(=O)NC=C1C(=O)O

Specifications

Melting Point 314°C to 316°C
Quantity 100 g
Beilstein 472182
Formula Weight 139.11
Percent Purity 98%
Chemical Name or Material 6-Hydroxynicotinic acid

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.