missing translation for 'onlineSavingsMsg'
Learn More

6-Hydroxynicotinic acid, 98+%

Product Code. 10193680 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
5 g
25 g
100 g
Packaging:
Glass bottle
Plastic bottle
missing translation for 'unitSize'
100g
25g
5g
This item is not returnable. View return policy

Product Code. 10193680

missing translation for 'mfr': Thermo Scientific Acros 156401000

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 5006-66-6 | C6H5NO3 | 139.11 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 5006-66-6
Molecular Formula C6H5NO3
Molecular Weight (g/mol) 139.11
MDL Number MFCD00006277
InChI Key BLHCMGRVFXRYRN-UHFFFAOYSA-N
Synonym 6-hydroxynicotinic acid, 2-hydroxy-5-pyridinecarboxylic acid, 6-hydroxypyridine-3-carboxylic acid, 6-hydroxy nicotinic acid, 3-pyridinecarboxylic acid, 1,6-dihydro-6-oxo, 2-hydroxypyridine-5-carboxylic acid, 2-pyridone-5-carboxylic acid, 6-hydroxy-nicotinic acid, 6-hydroxynicotinate, 1,6-dihydro-6-oxo-3-pyridinecarboxylic acid
PubChem CID 72924
ChEBI CHEBI:16168
IUPAC Name 6-oxo-1H-pyridine-3-carboxylic acid
SMILES C1=CC(=O)NC=C1C(=O)O

Specifications

CAS Min % 98.5
CAS Max % 100.0
Melting Point 314°C to 316°C
Color White to Brown
Infrared Spectrum Authentic
Assay Percent Range 99%
Packaging Plastic bottle
Quantity 100 g
Beilstein 22, 215
Formula Weight 139.11
Percent Purity 99%
Physical Form Powder
Chemical Name or Material 6-Hydroxynicotinic acid, 99%
Show More Show Less
compliance-icons
Product Identifier
  • 6-Hydroxynicotinic acid
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.