missing translation for 'onlineSavingsMsg'
Learn More
Learn More
6-Hydroxymethylpterin, Mikromol™
Click to view available options
Quantity:
50 mg
Unit Size:
50mg
Description
6-Hydroxymethylpterin
Specifications
Specifications
| Chemical Name or Material | 6-Hydroxymethylpterin |
| CAS | 712-29-8 |
| Analyte or Component Names | 6-Hydroxymethylpterin |
| Shipping Condition | Room Temperature |
| Molecular Formula | C7 H7 N5 O2 |
| Certifications/Compliance | ISO 9001 |
| Synonym | 6-Hydroxymethylpterin |
| InChI Formula | InChI=1S/C7H7N5O2/c8-7-11-5-4(6(14)12-7)10-3(2-13)1-9-5/h1,13H,2H2,(H3,8,9,11,12,14) |
| SMILES | NC1=NC(=O)c2nc(CO)cnc2N1 |
| IUPAC Name | 2-amino-6-(hydroxymethyl)-1H-pteridin-4-one |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction