missing translation for 'onlineSavingsMsg'
Learn More

6-Hydroxymelatonin Glucuronide, TRC

Product Code. 30390915
Change view
Click to view available options
Quantity:
1 mg
5 mg
10 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30390915 5 mg
30398246 10 mg
30407494 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30390915 Supplier Toronto Research Chemicals Supplier No. TRCH9446205MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 94840-69-4
Molecular Formula C19H24N2O9
Molecular Weight (g/mol) 424.4
Synonym 3-[2-(Acetylamino)ethyl]-5-methoxy-1H-indol-6-yl beta-D-Glucopyranosiduronic Acid
IUPAC Name (2S,3S,4S,5R,6S)-6-[[3-(2-acetamidoethyl)-5-methoxy-1H-indol-6-yl]oxy]-3,4,5-trihydroxyoxane-2-carboxylic acid
SMILES COc1cc2c(CCNC(=O)C)c[nH]c2cc1O[C@@H]3O[C@@H]([C@@H](O)[C@H](O)[C@H]3O)C(=O)O

Specifications

Quantity 5 mg
InChI Formula InChI=1S/C19H24N2O9/c1-8(22)20-4-3-9-7-21-11-6-13(12(28-2)5-10(9)11)29-19-16(25)14(23)15(24)17(30-19)18(26)27/h5-7,14-17,19,21,23-25H,3-4H2,1-2H3,(H,20,22)(H,26,27)/t14-,15-,16+,17-,19+/m0/s1
Formula Weight 424.1482
Chemical Name or Material 6-Hydroxymelatonin Glucuronide
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.