missing translation for 'onlineSavingsMsg'
Learn More

6α-Hydroxymedroxyprogesterone Acetate, Mikromol

Product Code. 30457617
Change view
Click to view available options
Quantity:
25 mg
Unit Size:
25mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30457617 25 mg 25mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30457617 Supplier Mikromol Supplier No. MM0532.080025
 Restricted Item
Estimated Shipment: 30-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Discover Mikromol- your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.

6α-Hydroxymedroxyprogesterone Acetate, Mikromol
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 6alpha-Hydroxy Medroxy Progesterone 17-Acetate
CAS 984-46-3
Analyte or Component Names 6alpha-Hydroxy Medroxy Progesterone 17-Acetate
Shipping Condition Room Temperature
Molecular Formula C24 H34 O5
Certifications/Compliance ISO 9001
Synonym Pregn-4-ene-3,20-dione, 17-(acetyloxy)-6-hydroxy-6-methyl-, (6α)- (9CI), Pregn-4-ene-3,20-dione, 6α,17-dihydroxy-6-methyl-, 17-acetate (7CI,8CI), 6α-Hydroxymedroxyprogesterone acetate, 6α-Hydroxymedroxyprogesterone 17-acetate
InChI Formula InChI=1S/C24H34O5/c1-14(25)24(29-15(2)26)11-8-19-17-13-23(5,28)20-12-16(27)6-9-21(20,3)18(17)7-10-22(19,24)4/h12,17-19,28H,6-11,13H2,1-5H3/t17-,18+,19+,21-,22+,23+,24+/m1/s1
Solution Type Neat
SMILES CC(=O)O[C@@]1(CC[C@H]2[C@@H]3C[C@](C)(O)C4=CC(=O)CC[C@]4(C)[C@H]3CC[C@]12C)C(=O)C
IUPAC Name [(6S,8R,9S,10R,13S,14S,17R)-17-acetyl-6-hydroxy-6,10,13-trimethyl-3-oxo-1,2,7,8,9,11,12,14,15,16-decahydrocyclopenta[a]phenanthren-17-yl] acetate
Molecular Weight (g/mol) 402.52
Quantity 25 mg
Formula Weight 402.2406 g/mol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.