missing translation for 'onlineSavingsMsg'
Learn More

6-Hydroxyindole, TRC

Product Code. 30399259
Change view
Click to view available options
Quantity:
500 mg
1 g
2.5 g
Unit Size:
1g
2.5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30399259 500 mg -
30402283 2.5 g 2.5g
30404812 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30399259 Supplier Toronto Research Chemicals Supplier No. TRCH953933500MG

Please to purchase this item. Need a web account? Register with us today!

This item requires a price update prior to being ordered.

Request a Price for this item here.

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2380-86-1
Molecular Formula C8 H7 N O
Molecular Weight (g/mol) 133.1473
Synonym Indol-6-ol (6CI,7CI,8CI), 6-Hydroxyindole
IUPAC Name 1H-indol-6-ol
SMILES Oc1ccc2cc[nH]c2c1

Specifications

Quantity 500 mg
InChI Formula InChI=1S/C8H7NO/c10-7-2-1-6-3-4-9-8(6)5-7/h1-5,9-10H
Formula Weight 133.0528
Chemical Name or Material 6-Hydroxyindole
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.