missing translation for 'onlineSavingsMsg'
Learn More

6-Hydroxyhexyl Methanethiosulfonate, TRC

Product Code. 30407467
Change view
Click to view available options
Quantity:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30407467 50 mg
30398408 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30407467 Supplier Toronto Research Chemicals Supplier No. TRCH94285050MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 212261-98-8
Molecular Formula C7 H16 O3 S2
Molecular Weight (g/mol) 212.33
IUPAC Name 6-methylsulfonylsulfanylhexan-1-ol
SMILES CS(=O)(=O)SCCCCCCO

Specifications

Quantity 50 mg
InChI Formula InChI=1S/C7H16O3S2/c1-12(9,10)11-7-5-3-2-4-6-8/h8H,2-7H2,1H3
Formula Weight 212.0541
Chemical Name or Material 6-Hydroxyhexyl Methanethiosulfonate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.