missing translation for 'onlineSavingsMsg'
Learn More

6-Hydroxy Warfarin Beta-D-Glucuronide D Disodium Salt Hydrate, TRC

Product Code. 30404640
Change view
Click to view available options
Quantity:
1 mg
10 mg
Unit Size:
10mg
1mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30404640 1 mg 1mg
30390019 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30404640 Supplier Toronto Research Chemicals Supplier No. TRCH9961101MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 6-Hydroxy Warfarin beta-D-Glucuronide D Disodium Salt Hydrate
Analyte or Component Names 6-Hydroxy Warfarin beta-D-Glucuronide D Disodium Salt Hydrate
Shipping Condition Room Temperature
Molecular Formula C25H22Na2O11 •xH2O
Synonym 4-Hydroxy-2-oxo-3-(3-oxo-1-phenylbutyl)-2H-1-benzopyran-6-ylβ-D-Glucopyranosiduronic Acid Disodium Salt Hydrate
InChI Formula InChI=1S/C25H24O11.2Na/c1-11(26)9-14(12-5-3-2-4-6-12)17-18(27)15-10-13(7-8-16(15)35-24(17)33)34-25-21(30)19(28)20(29)22(36-25)23(31)32;;/h2-8,10,14,19-22,25,27-30H,9H2,1H3,(H,31,32);;/q;2*+1/p-2/t14?,19-,20-,21+,22-,25+;;/m0../s1
Solution Type Neat
SMILES O=C(O1)C(C(C2=CC=CC=C2)CC(C)=O)=C(O[Na])C3=C1C=CC(O[C@@H]4O[C@H](C(O[Na])=O)[C@@H](O)[C@H](O)[C@H]4O)=C3
Industry Type Forensics
Molecular Weight (g/mol) 544.421802
Quantity 1 mg
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.