missing translation for 'onlineSavingsMsg'
Learn More

6’-Hydroxy Doxazosin, TRC

Product Code. 30396678
Change view
Click to view available options
Quantity:
50 mg
Unit Size:
50mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30396678 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30396678 Supplier Toronto Research Chemicals Supplier No. TRCH94176050MG
 Restricted Item
Estimated Shipment: 15-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 6'-Hydroxy Doxazosin
CAS 102932-29-6
Molecular Formula C23 H25 N5 O6
Quantity 50 mg
Synonym 6’-Hydroxy Doxazosin
InChI Formula InChI=1S/C23H25N5O6/c1-31-17-10-14-15(11-18(17)32-2)25-23(26-21(14)24)28-7-5-27(6-8-28)22(30)20-12-33-19-9-13(29)3-4-16(19)34-20/h3-4,9-11,20,29H,5-8,12H2,1-2H3,(H2,24,25,26)
SMILES COc1cc2nc(nc(N)c2cc1OC)N3CCN(CC3)C(=O)C4COc5cc(O)ccc5O4
IUPAC Name [4-(4-amino-6,7-dimethoxyquinazolin-2-yl)piperazin-1-yl]-(6-hydroxy-2,3-dihydro-1,4-benzodioxin-2-yl)methanone
Molecular Weight (g/mol) 467.47
Formula Weight 467.1805
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.