missing translation for 'onlineSavingsMsg'
Learn More

6-Hydroxy Chlorzoxazone-d2, TRC

Product Code. 30399553
Change view
Click to view available options
Quantity:
0.5 mg
5 mg
Unit Size:
0.5mg
5mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30399553 5 mg 5mg
30392591 0.5 mg 0.5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 30399553 Supplier Toronto Research Chemicals Supplier No. TRCH8251235MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 6-Hydroxy Chlorzoxazone-d2
CAS 1432065-00-3
Purity Grade Notes HPLC
Molecular Formula C7H2D2ClNO3
Quantity 5 mg
Synonym 5-Chloro-6-hydroxy-2(3H)-benz-d2-oxazolone, 6-Hydroxychlorzoxazone-d2, NSC 24955-d2
InChI Formula InChI=1S/C7H4ClNO3/c8-3-1-4-6(2-5(3)10)12-7(11)9-4/h1-2,10H,(H,9,11)/i1D,2D
SMILES OC1=C([2H])C2=C(NC(O2)=O)C([2H])=C1Cl
IUPAC Name 5-chloro-4,7-dideuterio-6-hydroxy-3H-1,3-benzoxazol-2-one
Molecular Weight (g/mol) 187.58
Formula Weight 187.0005
Percent Purity >95
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.