missing translation for 'onlineSavingsMsg'
Learn More

6-Hydroxy-2-naphthoic acid, 99%

Product Code. p-7026021
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
This item is not returnable. View return policy

Product Code. 11420447

Brand: Thermo Scientific Alfa Aesar B21791.03

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

CAS: 16712-64-4 | C11H8O3 | 188.182 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 16712-64-4
Molecular Formula C11H8O3
Molecular Weight (g/mol) 188.182
MDL Number MFCD00060070
InChI Key KAUQJMHLAFIZDU-UHFFFAOYSA-N
Synonym 6-hydroxy-2-naphthoic acid, 2-naphthalenecarboxylic acid, 6-hydroxy, 6-hydroxy-2-naphthalenecarboxylic acid, 6-carboxy-2-naphthol, 2-hydroxynaphthalene-6-carboxylic acid, 6-hydroxy-beta-naphthoic acid, unii-72xa858i6p, 6-hydroxy-naphthalene-2-carboxylic acid, 2-naphthoic acid, 6-hydroxy, 2-hydroxy-6-naphthoic acid
PubChem CID 85557
IUPAC Name 6-hydroxynaphthalene-2-carboxylic acid
SMILES C1=CC2=C(C=CC(=C2)O)C=C1C(=O)O

Specifications

Melting Point 248°C to 250°C
Quantity 1 g
Beilstein 2692084
Formula Weight 188.18
Percent Purity 99%
Chemical Name or Material 6-Hydroxy-2-naphthoic acid

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.