missing translation for 'onlineSavingsMsg'
Learn More

6-Hydroxy-2-naphthalenesulfonic Acid Sodium Salt, TRC

Product Code. 30396979
Change view
Click to view available options
Quantity:
25 g
Unit Size:
25g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30396979 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30396979 Supplier Toronto Research Chemicals Supplier No. TRCH95073525G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 6-Hydroxy-2-naphthalenesulfonic Acid Sodium Salt
CAS 135-76-2
Purity Grade Notes HPLC
Molecular Formula C10 H7 O4 S . Na
Quantity 25 g
Synonym 6-Hydroxy-2-naphthalenesulfonic Acid Sodium Salt (1:1), 2-Naphthalenesulfonic acid, 6-hydroxy-, monosodium salt (8CI,9CI), 2-Hydroxy-6-naphthalenesulfonic acid sodium salt, 2-Naphthol-6-sodium sulfonate, 2-Naphthol-6-sulfonic acid sodium salt, 6-Sulfo-2-naphthol sodium salt, Schaeffer salt, Schaeffer's salt, Sodium 2-naphthol-6-sulfonate, Sodium 6-hydroxy-2-naphthalenesulfonate, Sodium β-naphthol-6-sulfonate
InChI Formula InChI=1S/C10H8O4S.Na/c11-9-3-1-8-6-10(15(12,13)14)4-2-7(8)5-9;/h1-6,11H,(H,12,13,14);/q;+1/p-1
SMILES [Na+].Oc1ccc2cc(ccc2c1)S(=O)(=O)[O-]
IUPAC Name sodium;6-hydroxynaphthalene-2-sulfonate
Molecular Weight (g/mol) 246.22
Formula Weight 245.9963
Percent Purity >95
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.