missing translation for 'onlineSavingsMsg'
Learn More

6-Hydroxy-2,4,5-triaminopyrimidine, Sulfate, TRC

Product Code. 30401491
Change view
Click to view available options
Quantity:
1 g
2.5 g
25 g
Unit Size:
2.5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30401491 25 g -
30406723 2.5 g 2.5g
30397137 1 g -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30401491 Supplier Toronto Research Chemicals Supplier No. TRCH97125025G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 35011-47-3
Molecular Formula C4 H7 N5 O . H2 O . H2 O4 S
Molecular Weight (g/mol) 257.22
Synonym 4(3H)-Pyrimidinone, 2,5,6-triamino-, sulfate (1:1), 4(1H)-Pyrimidinone, 2,5,6-triamino-, sulfate (1:1) (9CI), NSC 167378, 2,5,6-Triaminopyrimidin-4(1H)-one monosulfate
IUPAC Name sulfuric acid;2,5,6-triaminopyrimidin-4-ol;hydrate
SMILES O.Nc1nc(N)c(N)c(O)n1.OS(=O)(=O)O

Specifications

Quantity 25 g
InChI Formula InChI=1S/C4H7N5O.H2O4S.H2O/c5-1-2(6)8-4(7)9-3(1)10;1-5(2,3)4;/h5H2,(H5,6,7,8,9,10);(H2,1,2,3,4);1H2
Formula Weight 257.043
Chemical Name or Material 6-Hydroxy-2,4,5-triaminopyrimidine, Sulfate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.