missing translation for 'onlineSavingsMsg'
Learn More

6-Hydroxy-1-tetralone, 98%, Thermo Scientific Chemicals

Product Code. 10735125 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10735125 1 g Glass bottle 1g
10224624 5 g Glass bottle 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10735125 Supplier Thermo Scientific Acros Supplier No. 398020010
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 3470-50-6
Molecular Formula C10H10O2
Molecular Weight (g/mol) 162.19
MDL Number MFCD00156670
InChI Key FNSQPQKPPGALFA-UHFFFAOYSA-N
Synonym 6-hydroxy-1-tetralone, 6-hydroxy-3,4-dihydronaphthalen-1 2h-one, 6-hydroxy-3,4-dihydro-1 2h-naphthalenone, 6-hydroxy-1,2,3,4-tetrahydronaphthalen-1-one, 6-hydroxytetralone, 1-tetralon-6-ol, 6-hydroxytetralin-1-one, 2,5-dimethoxy naphthalene, 1 2h-naphthalenone, 3,4-dihydro-6-hydroxy, 6-hydroxy-2,3,4-trihydronaphthalen-1-one
PubChem CID 4679494
IUPAC Name 6-hydroxy-3,4-dihydro-2H-naphthalen-1-one
SMILES C1CC2=C(C=CC(=C2)O)C(=O)C1

Specifications

CAS Min % 97.5
CAS Max % 100.0
Melting Point 155.0°C to 158.0°C
Color White to Brown
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (HPLC)
Packaging Glass bottle
Quantity 1 g
Formula Weight 162.19
Percent Purity 98%
Physical Form Crystalline Powder
Chemical Name or Material 6-Hydroxy-1-tetralone
Show More Show Less
compliance-icons
Product Identifier
  • 6-Hydroxy-1-tetralone
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P264-Wash thoroughly after handling.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
  • P332+P313-If skin irritation occurs: Get medical advice/attention.
  • P337+P313-If eye irritation persists: Get medical advice/attention.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.