missing translation for 'onlineSavingsMsg'
Learn More

6-Hydroxy-1-tetralone, 98%, Thermo Scientific Chemicals

Catalog Number p-3759060
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
This item is not returnable. View return policy
3470-50-6
C10H10O2
162.19
MFCD00156670
FNSQPQKPPGALFA-UHFFFAOYSA-N
6-hydroxy-1-tetralone, 6-hydroxy-3,4-dihydronaphthalen-1 2h-one, 6-hydroxy-3,4-dihydro-1 2h-naphthalenone, 6-hydroxy-1,2,3,4-tetrahydronaphthalen-1-one, 6-hydroxytetralone, 1-tetralon-6-ol, 6-hydroxytetralin-1-one, 2,5-dimethoxy naphthalene, 1 2h-naphthalenone, 3,4-dihydro-6-hydroxy, 6-hydroxy-2,3,4-trihydronaphthalen-1-one
4679494
6-hydroxy-3,4-dihydro-2H-naphthalen-1-one
C1CC2=C(C=CC(=C2)O)C(=O)C1
This item is not returnable. View return policy
3470-50-6
C10H10O2
162.19
MFCD00156670
FNSQPQKPPGALFA-UHFFFAOYSA-N
6-hydroxy-1-tetralone, 6-hydroxy-3,4-dihydronaphthalen-1 2h-one, 6-hydroxy-3,4-dihydro-1 2h-naphthalenone, 6-hydroxy-1,2,3,4-tetrahydronaphthalen-1-one, 6-hydroxytetralone, 1-tetralon-6-ol, 6-hydroxytetralin-1-one, 2,5-dimethoxy naphthalene, 1 2h-naphthalenone, 3,4-dihydro-6-hydroxy, 6-hydroxy-2,3,4-trihydronaphthalen-1-one
4679494
6-hydroxy-3,4-dihydro-2H-naphthalen-1-one
C1CC2=C(C=CC(=C2)O)C(=O)C1

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
CAS Min % 97.5
CAS Max % 100.0
Melting Point 155.0°C to 158.0°C
Color White to Brown
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (HPLC)
Packaging Glass bottle
Quantity 1 g
Formula Weight 162.19
Percent Purity 98%
Physical Form Crystalline Powder
Chemical Name or Material 6-Hydroxy-1-tetralone
Show More Show Less
compliance-icons
Product Identifier
  • 6-Hydroxy-1-tetralone
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P264-Wash thoroughly after handling.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
  • P332+P313-If skin irritation occurs: Get medical advice/attention.
  • P337+P313-If eye irritation persists: Get medical advice/attention.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'