missing translation for 'onlineSavingsMsg'
Learn More

6-Hydroxy-1-tetralone, 97%

Product Code. 15465607
Change view
Click to view available options
Quantity:
5 g
Unit Size:
5g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15465607 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15465607 Supplier Thermo Scientific Alfa Aesar Supplier No. H32840.06
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 3470-50-6 | C10H10O2 | 162.188 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Specifications

CAS 3470-50-6
Melting Point 154°C to 157°C
Molecular Formula C10H10O2
MDL Number MFCD00156670
Quantity 5 g
Synonym 6-hydroxy-1-tetralone, 6-hydroxy-3,4-dihydronaphthalen-1 2h-one, 6-hydroxy-3,4-dihydro-1 2h-naphthalenone, 6-hydroxy-1,2,3,4-tetrahydronaphthalen-1-one, 6-hydroxytetralone, 1-tetralon-6-ol, 6-hydroxytetralin-1-one, 2,5-dimethoxy naphthalene, 1 2h-naphthalenone, 3,4-dihydro-6-hydroxy, 6-hydroxy-2,3,4-trihydronaphthalen-1-one
InChI Key FNSQPQKPPGALFA-UHFFFAOYSA-N
SMILES C1CC2=C(C=CC(=C2)O)C(=O)C1
IUPAC Name 6-hydroxy-3,4-dihydro-2H-naphthalen-1-one
Molecular Weight (g/mol) 162.188
PubChem CID 4679494
Formula Weight 162.19
Percent Purity 97%
Chemical Name or Material 6-Hydroxy-1-tetralone
Show More Show Less
compliance-icons
Product Identifier
  • 6-Hydroxy-1-tetralone
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P264-Wash face, hands and anyexposed skin thoroughly after handling.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
  • P332+P313-If skin irritation occurs: Get medical advice/attention.
  • P337+P313-If eye irritation persists: Get medical advice/attention.
Supplemental information
  • MIXTURE LIST-Contain: 6-Hydroxy-1-tetralone

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.