missing translation for 'onlineSavingsMsg'
Learn More

6-Hydrazino-1,3,4-trimethyl-1H-pyrazolo[3,4-b]pyridine, 97%, Thermo Scientific™

Product Code. 10593032
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
Unit Size:
1g
250mg
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
10593032 1 g 1g
10672633 250 mg 250mg
10798625 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10593032 Supplier Thermo Scientific Maybridge Supplier No. KM02421DA

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Chemical Identifiers

CAS 175202-00-3
Molecular Formula C9H13N5
Molecular Weight (g/mol) 191.24
MDL Number MFCD00067969
InChI Key NDFXPLOECYNSHX-UHFFFAOYSA-N
Synonym 6-hydrazino-1,3,4-trimethyl-1h-pyrazolo 3,4-b pyridine, 6-hydrazinyl-1,3,4-trimethyl-1h-pyrazolo 3,4-b pyridine, 6-hydrazinyl-1,3,4-trimethylpyrazolo 3,4-b pyridine, 1,3,4-trimethylpyrazolo 5,4-b pyridine-6-ylhydrazine, 1,3,4-trimethylpyrazolo 3,4-b pyridin-6-yl hydrazine, 1h-pyrazolo 3,4-b pyridine,6-hydrazinyl-1,3,4-trimethyl, 6h-pyrazolo 3,4-b pyridin-6-one, 1,7-dihydro-1,3,4-trimethyl-, hydrazone 9ci
PubChem CID 2778646
IUPAC Name 6-hydrazinyl-1,3,4-trimethyl-1H-pyrazolo[3,4-b]pyridine
SMILES CN1N=C(C)C2=C(C)C=C(NN)N=C12

Specifications

CAS Min % 97.0
Assay Percent Range 97%
Packaging Amber glass bottle
Quantity 1 g
Formula Weight 191.24
Percent Purity 97%
Chemical Name or Material 6-Hydrazino-1,3,4-trimethyl-1H-pyrazolo[3,4-b]pyridine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.