missing translation for 'onlineSavingsMsg'
Learn More

6-Heptynoic Acid, TRC

Product Code. 30399640
Change view
Click to view available options
Quantity:
500 mg
1 g
5 g
Unit Size:
1g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30399640 500 mg -
30389615 5 g -
30398700 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30399640 Supplier Toronto Research Chemicals Supplier No. TRCH265710500MG

Please to purchase this item. Need a web account? Register with us today!

This item requires a price update prior to being ordered.

Request a Price for this item here.

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 30964-00-2
Molecular Formula C7H10O2
Molecular Weight (g/mol) 126.15
IUPAC Name hept-6-ynoic acid
SMILES OC(=O)CCCCC#C

Specifications

Quantity 500 mg
InChI Formula InChI=1S/C7H10O2/c1-2-3-4-5-6-7(8)9/h1H,3-6H2,(H,8,9)
Formula Weight 126.0681
Chemical Name or Material 6-Heptynoic Acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.