missing translation for 'onlineSavingsMsg'
Learn More

6-Heptynoic acid, 95%

Product Code. 15490428
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15490428 25 g 25g
15470428 1 g 1g
15480428 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 15490428 Supplier Thermo Scientific Alfa Aesar Supplier No. H53519.14
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 30964-00-2 | C7H10O2 | 126.16 g/mol

6-Heptynoic acid is used as a precursor for the the preparation of boradiazaindacenes (BODIPY dyes) by reacting with oxalylchloirde. It is used to prepare 4,4-difluoro-8-(hept-6-yne)-3,5-di(2-(4-methoxyphenyl))-4-bora-3a,4a-diaza-sindacene by reacting with 2-(4-methoxyphenyl)-1H-pyrrole.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
6-Heptynoic acid is used as a precursor for the the preparation of boradiazaindacenes (BODIPY dyes) by reacting with oxalylchloirde. It is used to prepare 4,4-difluoro-8-(hept-6-yne)-3,5-di(2-(4-methoxyphenyl))-4-bora-3a,4a-diaza-sindacene by reacting with 2-(4-methoxyphenyl)-1H-pyrrole.

Solubility
Miscible with dimethylformamide.

Notes
Incompatible with oxidizing agents and bases
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 30964-00-2
Molecular Formula C7H10O2
Molecular Weight (g/mol) 126.16
MDL Number MFCD00239430
InChI Key OFCPMJGTZUVUSM-UHFFFAOYSA-N
Synonym 6-heptynoic acid, 6-heptynoicacid, 6-heptynoic alcohol, ksc222a4l, ofcpmjgtzuvusm-uhfffaoysa
PubChem CID 4377950
IUPAC Name hept-6-ynoic acid
SMILES OC(=O)CCCCC#C

Specifications

Density 0.997
Boiling Point 93°C to 94°C (1 mmHg)
Flash Point 113°C (235°F)
Linear Formula HC≡C(CH2)4CO2H
Refractive Index 1.451
Quantity 25 g
UN Number UN3265
Beilstein 1747024
Solubility Information Miscible with dimethylformamide.
Formula Weight 126.16
Percent Purity 95%
Chemical Name or Material 6-Heptynoic acid
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.