missing translation for 'onlineSavingsMsg'
Learn More
Learn More
6-Formyl-2-(methylsulfanyl)nicotinonitrile, 97%, Thermo Scientific™
Click to view available options
Quantity:
250 mg
1 g
5 g
Packaging:
Amber Glass Bottle
Specifications
Specifications
Assay Percent Range | 97% |
Packaging | Amber Glass Bottle |
Quantity | 1 g |
Formula Weight | 178.21 |
Percent Purity | 97% |
Chemical Name or Material | 6-Formyl-2-(methylsulfanyl)nicotinonitrile |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'