missing translation for 'onlineSavingsMsg'
Learn More

6-Formyl-2-(methylsulfanyl)nicotinonitrile, 97%, Thermo Scientific™

Product Code. 10103624
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
Packaging:
Amber Glass Bottle
Unit Size:
1g
250mg
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10103624 5 g Amber Glass Bottle 5g
10039531 250 mg Amber Glass Bottle 250mg
10495522 1 g Amber Glass Bottle 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10103624 Supplier Thermo Scientific Maybridge Supplier No. SPB04394DE
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Chemical Identifiers

CAS 175277-27-7
Molecular Formula C8H6N2OS
Molecular Weight (g/mol) 178.21
MDL Number MFCD00173824
InChI Key UBGVOMZGBKLUOU-UHFFFAOYSA-N
Synonym 6-formyl-2-methylsulfanyl nicotinonitrile, 3-cyano-2-methylthio pyridine-6-carboxaldehyde, 6-formyl-2-methylsulfanyl pyridine-3-carbonitrile, 6-formyl-2-methylthio nicotinonitrile, 6-formyl-2-methylthio pyridine-3-carbonitrile, 6-formyl-2-methylthio-3-pyridinecarbonitrile, 3-pyridinecarbonitrile,6-formyl-2-methylthio, 6-formyl-2-methylsulphanyl nicotinonitrile, 6-formyl-2-methylthiopyridine-3-carbonitrile, 6-formyl-2-methylsulfanyl-pyridine-3-carbonitrile
PubChem CID 2799504
IUPAC Name 6-formyl-2-methylsulfanylpyridine-3-carbonitrile
SMILES CSC1=C(C=CC(C=O)=N1)C#N

Specifications

Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 5 g
Formula Weight 178.21
Percent Purity 97%
Chemical Name or Material 6-Formyl-2-(methylsulfanyl)nicotinonitrile
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.