missing translation for 'onlineSavingsMsg'
Learn More

6-Fluorosalicylic acid, 98+%, Thermo Scientific™

Product Code. 10567814 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10567814 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10567814 Supplier Thermo Scientific Acros Supplier No. 398770010
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Specifications

CAS 67531-86-6
Melting Point 162°C to 164°C
Assay Percent Range 98% min. (HPLC)
Packaging Glass Bottle
Molecular Formula C7H5FO3
Linear Formula FC6H3(OH)COOH
Quantity 1 g
Synonym 6-fluorosalicylic acid, 6-fluoro-2-hydroxybenzoic acid, 2-fluoro-6-hydroxybenzoicacid, 6-fluorosalicylicacid, benzoic acid, 2-fluoro-6-hydroxy, 2-fluoro-6-hydoxybenzoic acid, pubchem4958, acmc-1baav, 2-carboxy-3-fluorophenol
InChI Key BCEKGWWLVKXZKK-UHFFFAOYSA-N
SMILES C1=CC(=C(C(=C1)F)C(=O)O)O
IUPAC Name 2-fluoro-6-hydroxybenzoic acid
Molecular Weight (g/mol) 156.11
PubChem CID 586777
Formula Weight 156.11
Percent Purity 98+%
Chemical Name or Material 6-Fluorosalicylic acid
Show More Show Less
compliance-icons
Product Identifier
  • 6-Fluorosalicylic acid
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H318-Causes serious eye damage.
  • H315-Causes skin irritation.
  • H335-May cause respiratory irritation.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.