missing translation for 'onlineSavingsMsg'
Learn More

6-Fluoroindole-3-carboxaldehyde, 98%, Thermo Scientific Chemicals

Catalog Number 15418228
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy
2795-41-7
C9H6FNO
163.15
MFCD00069703
CWCYUOSLRVAKQZ-UHFFFAOYSA-N
6-fluoroindole-3-carboxaldehyde, 6-fluoroindole-3-carbaldehyde, 6-fluoroindole-3-aldehyde, 6-fluoro-1h-indole-3-carboxaldehyde, 1h-indole-3-carboxaldehyde, 6-fluoro, 1h-6-fluoroindole-3-carboxaldehyde, pubchem7269, ksc495g4p, 6-fluoro-3-formyl-1h-indole, 6-fluorooindole-3-carboxaldehyde
262903
6-fluoro-1H-indole-3-carbaldehyde
FC1=CC=C2C(NC=C2C=O)=C1
This item is not returnable. View return policy
2795-41-7
C9H6FNO
163.15
MFCD00069703
CWCYUOSLRVAKQZ-UHFFFAOYSA-N
6-fluoroindole-3-carboxaldehyde, 6-fluoroindole-3-carbaldehyde, 6-fluoroindole-3-aldehyde, 6-fluoro-1h-indole-3-carboxaldehyde, 1h-indole-3-carboxaldehyde, 6-fluoro, 1h-6-fluoroindole-3-carboxaldehyde, pubchem7269, ksc495g4p, 6-fluoro-3-formyl-1h-indole, 6-fluorooindole-3-carboxaldehyde
262903
6-fluoro-1H-indole-3-carbaldehyde
FC1=CC=C2C(NC=C2C=O)=C1

6-Fluoroindole-3-carboxaldehyde is an important raw material and intermediate used in organic synthesis, pharmaceuticals and agrochemicals.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
6-Fluoroindole-3-carboxaldehyde is an important raw material and intermediate used in organic synthesis, pharmaceuticals and agrochemicals.

Solubility
Soluble in methanol.

Notes
Store in freezer(-20°C). Keep container tightly closed. Store in a cool, dry and well-ventilated place.
TRUSTED_SUSTAINABILITY
Color Yellow
Quantity 1 g
Solubility Information Soluble in methanol.
Formula Weight 163.15
Percent Purity 98%
Physical Form Crystalline Solid
Chemical Name or Material 6-Fluoroindole-3-carboxaldehyde

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'