missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals 6-Fluoro-DL-tryptophan, 97%

Product Code. 10675382 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
100 mg
500 mg
Packaging:
Glass bottle
Unit Size:
100mg
500mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10675382 100 mg Glass bottle 100mg
10677494 500 mg Glass bottle 500mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10675382 Supplier Thermo Scientific Chemicals Supplier No. 146741000
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

CAS: 7730-20-3 | C11H11FN2O2 | 222.22 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7730-20-3
Molecular Formula C11H11FN2O2
Molecular Weight (g/mol) 222.22
MDL Number MFCD00005653
InChI Key YMEXGEAJNZRQEH-UHFFFAOYSA-N
Synonym 6-fluoro-dl-tryptophan, 6-fluorotryptophan, 2-amino-3-6-fluoro-1h-indol-3-yl propanoic acid, dl-tryptophan, 6-fluoro, dl-6-fluorotryptophane, +--6-fluorotryptophan, tryptophan, 6-fluoro-, dl, 6-fluoro-d,l-tryptophan, tryptophan, 6-fluoro, 2-amino-3-6-fluoroindol-3-yl propanoic acid
PubChem CID 94937
IUPAC Name 2-amino-3-(6-fluoro-1H-indol-3-yl)propanoic acid
SMILES NC(CC1=CNC2=CC(F)=CC=C12)C(O)=O

Specifications

CAS Min % 96.0
CAS Max % 100.0
Melting Point 280°C to 285°C
Color Beige to White
Infrared Spectrum Authentic
Assay Percent Range 97%
Packaging Glass bottle
Quantity 100 mg
Formula Weight 222.22
Percent Purity 97%
Physical Form Powder
Chemical Name or Material 6-Fluoro-DL-tryptophan
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.