missing translation for 'onlineSavingsMsg'
Learn More

6-Fluoro-3-pyridinylboronic acid, 97%, Thermo Scientific™

Product Code. 10674735 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10674735 5 g Glass bottle 5g
10031304 1 g Glass bottle 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10674735 Supplier Thermo Scientific Acros Supplier No. 397160050

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Chemical Identifiers

CAS 351019-18-6
Molecular Formula C5H5BFNO2
Molecular Weight (g/mol) 140.91
MDL Number MFCD03411559
InChI Key OJBYZWHAPXIJID-UHFFFAOYSA-N
Synonym 2-fluoropyridine-5-boronic acid, 2-fluoro-5-pyridylboronic acid, 6-fluoropyridin-3-yl boronic acid, 6-fluoropyridine-3-boronic acid, 2-fluoro-5-pyridineboronic acid, 2-fluoropyridin-5-ylboronic acid, 2-fluoropyridin-5-boronic acid, 6-fluoro-3-pyridinylboronic acid, 6-fluoropyridin-3-yl-3-boronic acid, 6-fluoro-3-pyridineboronic acid
PubChem CID 2783397
IUPAC Name (6-fluoropyridin-3-yl)boronic acid
SMILES OB(O)C1=CC=C(F)N=C1

Specifications

CAS Min % 96.0
CAS Max % 100.0
Melting Point 172.0°C to 176.0°C
Assay Percent Range 97%
Packaging Glass bottle
Quantity 5 g
Formula Weight 140.91
Percent Purity 97%
Chemical Name or Material 6-Fluoro-3-pyridinylboronic acid
compliance-icons
Product Identifier
  • 2-Fluoropyridine-5-boronic acid
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.