missing translation for 'onlineSavingsMsg'
Learn More

6-Fluoro-3-(1,2,3,6-tetrahydro-4-pyridyl)indole, 97%, Thermo Scientific Chemicals

Product Code. 15418586
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
Unit Size:
1g
250mg
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15418586 1 g 1g
15438586 250 mg 250mg
15428586 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 15418586 Supplier Thermo Scientific Alfa Aesar Supplier No. H63920.03
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 180161-14-2
Molecular Formula C13H13FN2
Molecular Weight (g/mol) 216.26
MDL Number MFCD04114772
InChI Key FBWKDFSEFVFKFY-UHFFFAOYSA-N
Synonym 6-fluoro-3-1,2,3,6-tetrahydropyridin-4-yl-1h-indole, 6-fluoro-3-1,2,3,6-tetrahydro-pyridin-4-yl-1h-indole, 6-fluoro-3-1,2,3,6-tetrahydro-4-pyridinyl-1h-indole, 6-fluoro-3-4-1,2,5,6-tetrahydropyridyl indole, 4-6-fluoroindol-3-yl-1,2,5,6-tetrahydropyridine, 6-fluoro-3-1,2,3,6-tetrahydro4-pyridinyl-1h-indole, 3-1,2,3,6-tetrahydropyridine-4-yl-6-fluoro-1h-indole, 4-6-fluoro-1h-indol-3-yl-1,2,3,6-tetrahydropyridine
PubChem CID 11053072
IUPAC Name 6-fluoro-3-(1,2,3,6-tetrahydropyridin-4-yl)-1H-indole
SMILES FC1=CC=C2C(NC=C2C2=CCNCC2)=C1

Specifications

Color Yellow to Brown
Quantity 1 g
Beilstein 8627107
Formula Weight 216.25
Percent Purity 97%
Physical Form Solid
Chemical Name or Material 6-Fluoro-3-(1,2,3,6-tetrahydro-4-pyridyl)indole

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.