missing translation for 'onlineSavingsMsg'
Learn More

6-Fluoro-2-methylquinoline, 98+%

Product Code. 11430353
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11430353 5 g 5g
11440353 25 g 25g
11450353 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11430353 Supplier Thermo Scientific Alfa Aesar Supplier No. A11469.06
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 1128-61-6 | C10H8FN | 161.179 g/mol

Metal-free transfer hydrogenation of 6-fluoro-2-methylquinoline by employing several chiral Bronsted acid catalysts was carried out in combination with dihydropyridines as the hydride source in asymmetric metal-free synthesis of fluoroquinolones by organocatalytic hydrogenation. Hydrogenation product of 6-fluoro-2-methylquinoline is the key intermediate of the antibacterial agent.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Metal-free transfer hydrogenation of 6-fluoro-2-methylquinoline by employing several chiral Brønsted acid catalysts was carried out in combination with dihydropyridines as the hydride source in asymmetric metal-free synthesis of fluoroquinolones by organocatalytic hydrogenation. Hydrogenation product of 6-fluoro-2-methylquinoline is the key intermediate of the antibacterial agent.

Solubility
Soluble in Chloroform.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1128-61-6
Molecular Formula C10H8FN
Molecular Weight (g/mol) 161.179
MDL Number MFCD00041233
InChI Key GPIARMSVZOEZCV-UHFFFAOYSA-N
Synonym 6-fluoroquinaldine, quinoline, 6-fluoro-2-methyl, 2-methyl-6-fluoroquinoline, 6-fluor-2-methylchinolin, 6-fluoro quinaldine, pubchem7579, acmc-1boyk, maybridge1_000571, 6-fluoro-2-methyl-quinoline, 6-fluoro-2-methyl quinoline
PubChem CID 70784
IUPAC Name 6-fluoro-2-methylquinoline
SMILES CC1=NC2=C(C=C1)C=C(C=C2)F

Specifications

Melting Point 52°C to 56°C
Quantity 5 g
Beilstein 1281677
Solubility Information Soluble in Chloroform.
Formula Weight 161.18
Percent Purity ≥98%
Chemical Name or Material 6-Fluoro-2-methylquinoline

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.