missing translation for 'onlineSavingsMsg'
Learn More

6-Fluoro-2,3-dihydro-4H-thiochromen-4-one, ≥95%, Thermo Scientific™

Product Code. 10374632
Change view
Click to view available options
Quantity:
1 g
5 g
10 g
Unit Size:
10g
1g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10374632 10 g 10g
10223432 1 g 1g
10457944 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10374632 Supplier Thermo Scientific Maybridge Supplier No. SEW05178EA
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Chemical Identifiers

CAS 21243-18-5
Molecular Formula C9H7FOS
Molecular Weight (g/mol) 182.212
MDL Number MFCD00078488
InChI Key NBBGHADNMPMHST-UHFFFAOYSA-N
Synonym 6-fluorothio-4-chromanone, 6-fluoro-2,3-dihydro-4h-thiochromen-4-one, 6-fluorothiochroman-4-one, 6-fluoro-2,3-dihydro-4h-1-benzothiopyran-4-one, 6-fluoro-3,4-dihydro-2h-1-benzothiopyran-4-one, 4h-1-benzothiopyran-4-one,6-fluoro-2,3-dihydro, 6-fluoro-3,4-dihydro-2h-1-benzothiin-4-one, 6-fluoro-2,3-dihydro-1-benzothiopyran-4-one, maybridge1_007680, 6-fluoro-4-oxothiochroman
PubChem CID 2737521
IUPAC Name 6-fluoro-2,3-dihydrothiochromen-4-one
SMILES C1CSC2=C(C1=O)C=C(C=C2)F

Specifications

CAS Min % 95.0
Assay Percent Range 95%
Packaging Amber glass bottle
Quantity 10 g
Formula Weight 182.22
Percent Purity 95%
Chemical Name or Material 6-Fluoro-2, 3-dihydro-4H-thiochromen-4-one
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.