missing translation for 'onlineSavingsMsg'
Learn More
Learn More
6-Fluoro-2,3-dihydro-4H-thiochromen-4-one, ≥95%, Thermo Scientific™
Click to view available options
Quantity:
1 g
5 g
10 g
Specifications
CAS Min % | 95.0 |
Assay Percent Range | 95% |
Packaging | Amber glass bottle |
Quantity | 1 g |
Formula Weight | 182.22 |
Percent Purity | 95% |
Chemical Name or Material | 6-Fluoro-2, 3-dihydro-4H-thiochromen-4-one |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'