missing translation for 'onlineSavingsMsg'
Learn More

6-Fluoro-1,2,3,4-tetrahydroquinoline, 97%

Product Code. 15437507
Change view
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15437507 250 mg 250mg
15427507 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15437507 Supplier Thermo Scientific Alfa Aesar Supplier No. H35364.MD
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 59611-52-8 | C9H10FN | 151.18 g/mol

6-Fluoro-1,2,3,4-tetrahydroquinoline is used as pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
6-Fluoro-1,2,3,4-tetrahydroquinoline is used as pharmaceutical intermediate.

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 59611-52-8
Molecular Formula C9H10FN
Molecular Weight (g/mol) 151.18
MDL Number MFCD09040997
InChI Key NECDDBBJVCNJNS-UHFFFAOYSA-N
Synonym quinoline, 6-fluoro-1,2,3,4-tetrahydro, 6-fluoro-1,2,3,4-tetrahydro-quinoline
PubChem CID 14783911
IUPAC Name 6-fluoro-1,2,3,4-tetrahydroquinoline
SMILES FC1=CC2=C(NCCC2)C=C1

Specifications

Melting Point 32°C to 34°C
Quantity 250 mg
Formula Weight 151.18
Percent Purity 97%
Chemical Name or Material 6-Fluoro-1,2,3,4-tetrahydroquinoline

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.