missing translation for 'onlineSavingsMsg'
Learn More

6-(Ethylamino)pyridine-3-carbonitrile, TRC

Product Code. 30388197
Change view
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30388197 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30388197 Supplier Toronto Research Chemicals Supplier No. TRCE899645100MG
 Restricted Item
Estimated Shipment: 14-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

CAS 1016813-34-5
Molecular Formula C8H9N3
Quantity 100 mg
Synonym 6-(Ethylamino)-3-pyridinecarbonitrile
InChI Formula InChI=1S/C8H9N3/c1-2-10-8-4-3-7(5-9)6-11-8/h3-4,6H,2H2,1H3,(H,10,11)
SMILES CCNc1ccc(cn1)C#N
Molecular Weight (g/mol) 147.18
Formula Weight 147.0796
Chemical Name or Material 6-(Ethylamino)pyridine-3-carbonitrile
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.