missing translation for 'onlineSavingsMsg'
Learn More

6-Ethyl-4-(1-piperazinyl)thieno[2,3-d]pyrimidine Hydrochloride, TRC

Product Code. 30387665
Change view
Click to view available options
Quantity:
250 mg
Unit Size:
250mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30387665 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30387665 Supplier Toronto Research Chemicals Supplier No. TRCE898795250MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 6-Ethyl-4-(1-piperazinyl)thieno[2,3-d]pyrimidine Hydrochloride
CAS 502648-93-3
Molecular Formula C12H16N4S • 2(HCl)
Quantity 250 mg
InChI Formula InChI=1S/C12H16N4S.2ClH/c1-2-9-7-10-11(14-8-15-12(10)17-9)16-5-3-13-4-6-16;;/h7-8,13H,2-6H2,1H3;2*1H
SMILES CCC1=CC2=C(S1)N=CN=C2N3CCNCC3.Cl
IUPAC Name 6-ethyl-4-(piperazin-1-yl)thieno[2,3-d]pyrimidine dihydrogen chloride
Molecular Weight (g/mol) 321.27
Formula Weight 320.07
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.