missing translation for 'onlineSavingsMsg'
Learn More

6-Ethoxy-2-naphthaleneboronic acid, 97%, Thermo Scientific™

Product Code. 11976011 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11976011 1 g 1g
11996011 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11976011 Supplier Thermo Scientific Acros Supplier No. 440230010

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Chemical Identifiers

CAS 352525-98-5
Molecular Formula C12H13BO3
Molecular Weight (g/mol) 216.04
MDL Number MFCD03427009
InChI Key INXXVGFSXYJGHI-UHFFFAOYSA-N
Synonym 6-ethoxy-2-naphthaleneboronic acid, 6-ethoxynaphthalen-2-yl boronic acid, 6-ethoxynaphthalene-2-boronic acid, boronic acid, 6-ethoxy-2-naphthalenyl, 2-ethoxy-6-naphthaleneboronic acid, boronic acid, b-6-ethoxy-2-naphthalenyl, 6-ethoxy-2-naphthalenyl-boronic acid, 6-ethoxy-2, acmc-209ies, boronic acid,b-6-ethoxy-2-naphthalenyl
PubChem CID 4641693
IUPAC Name (6-ethoxynaphthalen-2-yl)boronic acid
SMILES CCOC1=CC2=CC=C(C=C2C=C1)B(O)O

Specifications

CAS Min % 96.0
CAS Max % 100.0
Melting Point 238.5°C to 243.3°C
Color White
Infrared Spectrum Authentic
Assay Percent Range 96% min. (HPLC)
Packaging Glass Bottle
Quantity 1 g
Formula Weight 216.04
Percent Purity 97%
Physical Form Powder
Chemical Name or Material 6-Ethoxy-2-naphthaleneboronic acid
Show More Show Less
compliance-icons
Product Identifier
  • 6-Ethoxy-2-naphthaleneboronic acid
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.