missing translation for 'onlineSavingsMsg'
Learn More

6-Ethoxy-2-mercaptobenzothiazole, 99%

Product Code. 11331587
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11331587 5 g 5g
11341587 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11331587 Supplier Thermo Scientific Alfa Aesar Supplier No. L04447.06
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 120-53-6 | C9H9NOS2 | 211.297 g/mol

6-Ethoxy-2-mercaptobenzothiazole is used as a reagent in the preparation of nitrothiazolythio benzothiazoles as carbon-junction N-terminal kinase inhibitors. It is used in the synthesis of active pharmaceutical ingredients. Further, it serves as a matrix-assisted laser desorption mass spectrometry for bio molecules mass determination.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
6-Ethoxy-2-mercaptobenzothiazole is used as a reagent in the preparation of nitrothiazolythio benzothiazoles as carbon-junction N-terminal kinase inhibitors. It is used in the synthesis of active pharmaceutical ingredients. Further, it serves as a matrix-assisted laser desorption mass spectrometry for bio molecules mass determination.

Solubility
Insoluble in water.

Notes
Incompatible with strong oxidizing agents, strong bases and halogens.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 120-53-6
Molecular Formula C9H9NOS2
Molecular Weight (g/mol) 211.297
MDL Number MFCD00005782
InChI Key HOASVNMVYBSLSU-UHFFFAOYSA-N
Synonym 6-ethoxy-2-mercaptobenzothiazole, 6-ethoxybenzothiazole-2-thiol, 2 3h-benzothiazolethione, 6-ethoxy, 6-ethoxybenzothiazolethiol, usaf pd-58, 6-ethoxy-2-benzothiazolethiol, 6-ethoxybenzo d thiazole-2 3h-thione, 6-ethoxy-1,3-benzothiazole-2-thiol, 2-mercapto-6-ethoxybenzothiazole, 2-benzothiazolethiol, 6-ethoxy
PubChem CID 719344
IUPAC Name 6-ethoxy-3H-1,3-benzothiazole-2-thione
SMILES CCOC1=CC2=C(C=C1)NC(=S)S2

Specifications

Melting Point 196°C to 201°C
Quantity 5 g
Beilstein 152313
Solubility Information Insoluble in water.
Formula Weight 211.32
Percent Purity 99%
Chemical Name or Material 6-Ethoxy-2-mercaptobenzothiazole

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.