missing translation for 'onlineSavingsMsg'
Learn More

6-(Difluoromethoxy)-2-[[(3,4-dimethoxy-2-pyridinyl)methyl]sulfinyl]-1-methyl-1H-benzimidazole, TRC

Product Code. 30380624
Change view
Click to view available options
Quantity:
10 mg
25 mg
50 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30380624 50 mg
30387133 10 mg
30378161 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30380624 Supplier Toronto Research Chemicals Supplier No. TRCD45171850MG
 Restricted Item
Estimated Shipment: 03-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 721924-06-7
Molecular Formula C17H17F2N3O4S
Molecular Weight (g/mol) 397.4
Synonym Pantoprazole EP Impurity
SMILES COc1ccnc(CS(=O)c2nc3ccc(OC(F)F)cc3n2C)c1OC

Specifications

Quantity 50 mg
InChI Formula InChI=1S/C17H17F2N3O4S/c1-22-13-8-10(26-16(18)19)4-5-11(13)21-17(22)27(23)9-12-15(25-3)14(24-2)6-7-20-12/h4-8,16H,9H2,1-3H3
Formula Weight 397.0908
Chemical Name or Material 6-(Difluoromethoxy)-2-(((3,4-dimethoxy-2-pyridinyl)methyl)sulfinyl)-1-methyl-1H-benzimidazole
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.