missing translation for 'onlineSavingsMsg'
Learn More

6-(Diethylamino)-3-pyridinylaldehyde, 97%, Thermo Scientific™

Product Code. 10370643
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
Packaging:
Amber Glass Bottle
Unit Size:
1g
250mg
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10370643 250 mg Amber Glass Bottle 250mg
10681825 1 g Amber Glass Bottle 1g
10092444 5 g Amber Glass Bottle 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10370643 Supplier Thermo Scientific Maybridge Supplier No. MO07193CB
 Restricted Item
Estimated Shipment: 09-10-2026
Add to basket
This item is not returnable. View return policy

Chemical Identifiers

CAS 578726-67-7
Molecular Formula C10H14N2O
Molecular Weight (g/mol) 178.24
MDL Number MFCD06200894
InChI Key PCISUIMHLSRKGR-UHFFFAOYSA-N
Synonym 6-diethylamino pyridine-3-carbaldehyde, 6-diethylamino-3-pyridinylaldehyde, 6-diethylamino nicotinaldehyde, 6-diethylamino-3-pyridinecarboxaldehyde
PubChem CID 2794806
IUPAC Name 6-(diethylamino)pyridine-3-carbaldehyde
SMILES CCN(CC)C1=NC=C(C=O)C=C1

Specifications

CAS Min % 97.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 250 mg
Formula Weight 178.23
Percent Purity 97%
Chemical Name or Material 6-(Diethylamino)-3-pyridinylaldehyde
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.