missing translation for 'onlineSavingsMsg'
Learn More

[6-(Diethylamino)-3-pyridinyl]methanol, 97%, Thermo Scientific™

Product Code. 10225604
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
Unit Size:
1g
250mg
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10225604 5 g 5g
10670305 250 mg 250mg
10289742 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10225604 Supplier Thermo Scientific Maybridge Supplier No. MO07117DE
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Chemical Identifiers

CAS 690632-68-9
Molecular Formula C10H16N2O
Molecular Weight (g/mol) 180.251
MDL Number MFCD06200887
InChI Key OMGIPGLZKJOJSG-UHFFFAOYSA-N
Synonym 6-diethylamino-3-pyridinyl methanol, 6-diethylamino pyridin-3-yl methanol, 6-diethylamino-3-pyridyl methan-1-ol
PubChem CID 2794793
IUPAC Name [6-(diethylamino)pyridin-3-yl]methanol
SMILES CCN(CC)C1=NC=C(C=C1)CO

Specifications

CAS Min % 97.0
Assay Percent Range 97%
Quantity 5 g
Formula Weight 180.25
Percent Purity 97%
Chemical Name or Material [6-(Diethylamino)-3-pyridinyl]methanol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.