missing translation for 'onlineSavingsMsg'
Learn More

6-Dehydro Prednisolone, TRC

Product Code. 30361384
Change view
Click to view available options
Quantity:
10 mg
25 mg
50 mg
Unit Size:
10mg
25mg
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30361384 50 mg 50mg
30353054 25 mg 25mg
30357435 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30361384 Supplier Toronto Research Chemicals Supplier No. TRCD23000050MG
 Restricted Item
Estimated Shipment: 13-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2427-64-7
Molecular Formula C21H26O5
Molecular Weight (g/mol) 358.43
Synonym Pregna-1,4,6-triene-3,20-dione, 11,17,21-trihydroxy-, (11β)-, Pregna-1,4,6-triene-3,20-dione, 11β,17,21-trihydroxy- (6CI,7CI,8CI), (11β)-11,17,21-Trihydroxypregna-1,4,6-triene-3,20-dione, NSC 12882
SMILES C[C@]12C[C@H](O)[C@H]3[C@@H](C=CC4=CC(=O)C=C[C@]34C)[C@@H]1CC[C@]2(O)C(=O)CO

Specifications

Quantity 50 mg
InChI Formula InChI=1S/C21H26O5/c1-19-7-5-13(23)9-12(19)3-4-14-15-6-8-21(26,17(25)11-22)20(15,2)10-16(24)18(14)19/h3-5,7,9,14-16,18,22,24,26H,6,8,10-11H2,1-2H3/t14-,15-,16-,18+,19-,20-,21-/m0/s1
Formula Weight 358.178
Percent Purity >95%
Chemical Name or Material 6,7-Dehydro Prednisolone
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.