missing translation for 'onlineSavingsMsg'
Learn More

6-(Cyclopropylamino)-9-[(1R,4S)-4-[[(1,1-dimethylethyl)oxy]methyl]cyclopent-2-enyl]-9H-purine-2-amine (Abacavir tert-Butyl Ether), Mikromol™

Product Code. 30050481
Click to view available options
Quantity:
25 mg
Unit Size:
25mg
This item is not returnable. View return policy

Product Code. 30050481

Brand: Mikromol MM3249.110025

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.

6-(Cyclopropylamino)-9-[(1R,4S)-4-[[(1,1-dimethylethyl)oxy]methyl]cyclopent-2-enyl]-9H-purine-2-amine (Abacavir tert-Butyl Ether)
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Abacavir tert-Butyl Ether
CAS 1443421-68-8
Analyte or Component Names Abacavir tert-Butyl Ether
Shipping Condition Room Temperature
Molecular Formula C18 H26 N6 O
Certifications/Compliance ISO 9001
Synonym Abacavir tert-butyl ether, N6-Cyclopropyl-9-{(1R,4S)-4-[(1,1-dimethylethoxy)methyl]cyclopent-2-en-1-yl}-9H-purine-2,6-diamine, 6-(Cyclopropylamino)-9-[(1R,4S)-4-[[(1,1-dimethylethyl)oxy]methyl]cyclopent-2-enyl]-9H-purine-2-amine, 9H-Purine-2,6-diamine, N6-cyclopropyl-9-[(1R,4S)-4-[(1,1-dimethylethoxy)methyl]-2-cyclopenten-1-yl]-, N6-Cyclopropyl-9-[(1R,4S)-4-[(1,1-dimethylethoxy)methyl]-2-cyclopenten-1-yl]-9H-purine-2,6-diamine, Abacavir Sulfate Imp. F (Ph. Int.), Abacavir Sulfate Imp F (EP), Abacavir Sulfate Impurity F, Abacavir Impurity F
InChI Formula InChI=1S/C18H26N6O/c1-18(2,3)25-9-11-4-7-13(8-11)24-10-20-14-15(21-12-5-6-12)22-17(19)23-16(14)24/h4,7,10-13H,5-6,8-9H2,1-3H3,(H3,19,21,22,23)/t11-,13+/m1/s1
SMILES CC(C)(C)OC[C@H]1C[C@H](C=C1)n2cnc3c(NC4CC4)nc(N)nc23
IUPAC Name 6-N-cyclopropyl-9-[(1R,4S)-4-[(2-methylpropan-2-yl)oxymethyl]cyclopent-2-en-1-yl]purine-2,6-diamine
Molecular Weight (g/mol) 342.4386
Quantity 25 mg
Formula Weight 342.2168 g/mol
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.