missing translation for 'onlineSavingsMsg'
Learn More

6-Cyclohexyl-4-methyl-2H-pyran-2-one, TRC

Product Code. 30133019
Change view
Click to view available options
Quantity:
100 mg
1 g
Unit Size:
100mg
1g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30133019 1 g 1g
30003924 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30133019 Supplier Toronto Research Chemicals Supplier No. TRCC9880651G
 Restricted Item
Estimated Shipment: 17-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 14818-35-0
Molecular Formula C12 H16 O2
Molecular Weight (g/mol) 192.25
Synonym 2H-Pyran-2-one, 6-cyclohexyl-4-methyl-, 6-Cyclohexyl-4-methyl-2H-pyran-2-one, 6-Cyclohexyl-4-methyl-2-pyrone
IUPAC Name 6-cyclohexyl-4-methylpyran-2-one
SMILES CC1=CC(=O)OC(=C1)C2CCCCC2

Specifications

Quantity 1 g
InChI Formula InChI=1S/C12H16O2/c1-9-7-11(14-12(13)8-9)10-5-3-2-4-6-10/h7-8,10H,2-6H2,1H3
Formula Weight 192.115
Chemical Name or Material 6-cyclohexyl-4-methyl-2H-pyran-2-one
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.