missing translation for 'onlineSavingsMsg'
Learn More

6-Cyano-7-nitroquinoxaline-2,3-dione, TRC

Product Code. 30360253
Change view
Click to view available options
Quantity:
5 mg
25 mg
50 mg
Unit Size:
50mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30360253 5 mg 5mg
30357245 50 mg 50mg
30366011 25 mg -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30360253 Supplier Toronto Research Chemicals Supplier No. TRCC9820005MG
 Restricted Item
Estimated Shipment: 21-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 115066-14-3
Molecular Formula C9 H4 N4 O4
Molecular Weight (g/mol) 232.15
IUPAC Name 7-nitro-2,3-dioxo-1,4-dihydroquinoxaline-6-carbonitrile
SMILES [O-][N+](=O)c1cc2NC(=O)C(=O)Nc2cc1C#N

Specifications

Quantity 5 mg
InChI Formula InChI=1S/C9H4N4O4/c10-3-4-1-5-6(2-7(4)13(16)17)12-9(15)8(14)11-5/h1-2H,(H,11,14)(H,12,15)
Formula Weight 232.0233
Chemical Name or Material 6-Cyano-7-nitroquinoxaline-2,3-dione
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.