missing translation for 'onlineSavingsMsg'
Learn More

6-Cyano-7-nitro-1,4-dihydroquinoxaline-2,3-dione, 98%

Product Code. 15480307
Click to view available options
Quantity:
10 mg
25 mg
50 mg
Unit Size:
10mg
25mg
50mg
This item is not returnable. View return policy

Product Code. 15480307

Brand: Thermo Scientific Alfa Aesar J63472.MB

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Potent, selective kainate/quisqualate (non-NMDA) receptor antagonist | CAS: 115066-14-3 | C9H4N4O4 | 232.155 g/mol

Potent, selective kainate/quisqualate (non-NMDA) receptor antagonist

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 115066-14-3
Molecular Formula C9H4N4O4
Molecular Weight (g/mol) 232.155
MDL Number MFCD00069232
InChI Key RPXVIAFEQBNEAX-UHFFFAOYSA-N
Synonym cnqx, 6-cyano-7-nitroquinoxaline-2,3-dione, unii-6ote87sccw, 7-nitro-2,3-dioxo-1,2,3,4-tetrahydroquinoxaline-6-carbonitrile, 6ote87sccw, 3h cnqx, 6-cyano-2,3-dihydroxy-7-nitroquinoxaline, 1,4-dihydro-2,3-dihydroxy-7-nitro-6-quinoxalinecarbonitrile, 6-quinoxalinecarbonitrile, 1,2,3,4-tetrahydro-7-nitro-2,3-dioxo, 2,3-dihydroxy-7-nitro-6-quinoxalinecarbonitrile
PubChem CID 3721046
ChEBI CHEBI:34468
IUPAC Name 7-nitro-2,3-dioxo-1,4-dihydroquinoxaline-6-carbonitrile
SMILES C1=C(C(=CC2=C1NC(=O)C(=O)N2)[N+](=O)[O-])C#N

Specifications

Melting Point >300°C
Color Yellow
Quantity 25 mg
Beilstein 5821072
Solubility Information Soluble in DMSO to 100mM
Formula Weight 232.16
Percent Purity ≥99%
Physical Form Solid
Chemical Name or Material 6-Cyano-7-nitro-1,4-dihydroquinoxaline-2,3-dione

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.