missing translation for 'onlineSavingsMsg'
Learn More

6-chloropyrimidin-4(3H)-one, Thermo Scientific™

Product Code. 10091004
Change view
Click to view available options
Quantity:
1 g
5 g
Packaging:
Amber Glass Bottle
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10091004 5 g Amber Glass Bottle 5g
10106812 1 g Amber Glass Bottle 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10091004 Supplier Thermo Scientific Maybridge Supplier No. MO07170DE
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Chemical Identifiers

CAS 4765-77-9
Molecular Formula C4H3ClN2O
Molecular Weight (g/mol) 130.531
InChI Key AXFABVAPHSWFMD-UHFFFAOYSA-N
Synonym 6-chloro-4-hydroxypyrimidine, 6-chloropyrimidin-4-ol, 4-chloro-6-hydroxypyrimidine, 6-chloropyrimidin-4 1h-one, 6-chloropyrimidin-4 3h-one, 6-chloro-4-pyrimidinol, 6-chloro-4 1h-pyrimidinone, 4 1h-pyrimidinone, 6-chloro, 6-chloro-3,4-dihydro-4-oxopyrimidine, zlchem 18
PubChem CID 358318
IUPAC Name 6-chloro-1H-pyrimidin-4-one
SMILES C1=C(NC=NC1=O)Cl

Specifications

Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 5 g
Formula Weight 130.53
Percent Purity 97%
Chemical Name or Material 6-Chloro-4-pyrimidinol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.