missing translation for 'onlineSavingsMsg'
Learn More

6-Chloropyridine-3-boronic acid, 96%

Product Code. 11345659
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
Unit Size:
1g
250mg
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11345659 5 g 5g
11325659 250 mg 250mg
11335659 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11345659 Supplier Thermo Scientific Alfa Aesar Supplier No. L20388.06
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 444120-91-6 | C5H5BClNO2 | 157.36 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 444120-91-6
Molecular Formula C5H5BClNO2
Molecular Weight (g/mol) 157.36
MDL Number MFCD03094998
InChI Key WPAPNCXMYWRTTL-UHFFFAOYSA-N
Synonym 2-chloropyridine-5-boronic acid, 6-chloropyridin-3-yl boronic acid, 2-chloro-5-pyridineboronic acid, 2-chloro-5-pyridylboronic acid, 6-chloro-3-pyridylboronic acid, 6-chloropyridin-3-yl-3-boronic acid, 6-chloropyridine-3-boronic acid, 2-chloropyridin-5-ylboronic acid, 6-chloro-3-pyridinylboronic acid, 2-chloro-pyridine-5-boronic acid
PubChem CID 3845034
IUPAC Name (6-chloropyridin-3-yl)boronic acid
SMILES OB(O)C1=CC=C(Cl)N=C1

Specifications

Melting Point 154°C to 157°C
Quantity 5 g
Beilstein 9119810
Formula Weight 157.37
Percent Purity 96%
Chemical Name or Material 6-Chloropyridine-3-boronic acid

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.