missing translation for 'onlineSavingsMsg'
Learn More

6-chloropyridazine-3-carboxylic acid, 97%, Thermo Scientific™

Product Code. 10411538 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
Packaging:
Glass bottle
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10411538 1 g Glass bottle 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10411538 Supplier Thermo Scientific Acros Supplier No. 429530010
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Chemical Identifiers

CAS 5096-73-1
Molecular Formula C5H3ClN2O2
Molecular Weight (g/mol) 158.54
MDL Number MFCD00160464
InChI Key HHGZQZULOHYEOH-UHFFFAOYSA-N
Synonym 6-chloro-pyridazine-3-carboxylic acid, 3-chloropyridazine-6-carboxylic acid, 3-pyridazinecarboxylic acid, 6-chloro, 6-chloro-3-carboxy-pyridazine, 3-carboxy-6-chloropyridazine, 6-chloro-3-pyridazine-carboxylic acid, 6-chloropyridazine-3-carboxylic, 6-chloropyridazine-3-carboxylicacid, pubchem17692
PubChem CID 6415762
IUPAC Name 6-chloropyridazine-3-carboxylic acid
SMILES OC(=O)C1=NN=C(Cl)C=C1

Specifications

CAS Min % 96.0
CAS Max % 100.0
Melting Point 145.0°C to 148.0°C
Color White to Cream
Infrared Spectrum Authentic
Assay Percent Range 96% min. (HPLC)
Packaging Glass bottle
Quantity 1 g
Formula Weight 158.54
Percent Purity 97%
Physical Form Crystalline Powder
Chemical Name or Material 6-chloropyridazine-3-carboxylic acid
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.